Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009206.2 | NP_033232.2 | 6 | 1314 | ||
GenBank mRNA | AF035526.1 | - | 7 | 1197 | ||
AK138398.1 | - | 6 | 1269 | |||
AK147161.1 | - | 6 | 652 | |||
AK160250.1 | - | 5 | 1189 | |||
BC072588.1 | - | 6 | 1297 | |||
BC090964.1 | - | 6 | 866 | 30625511 |