Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011404.3 | NP_035534.2 | 3 | 780 | ||
GenBank mRNA | AB017189.1 | - | 3 | 726 | ||
AB023409.1 | - | 3 | 708 | |||
AK142288.1 | - | 3 | 781 | |||
AK148134.1 | - | 3 | 450 | |||
AK180682.1 | - | 2 | 96 | |||
AK183612.1 | - | 3 | 141 | |||
AK199678.1 | - | 2 | 96 | |||
AK203738.1 | - | 2 | 96 | |||
BC026131.1 | - | 3 | 789 | 4188854 |