Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291227.1 | NP_001278156.1 | 26 | 4519 | ||
NM_001291228.1 | NP_001278157.1 | 25 | 4495 | |||
NM_178804.4 | NP_848919.3 | 25 | 4483 | |||
GenBank mRNA | AF074960.1 | - | 11 | 1110 | ||
AF144628.1 | - | 25 | 2844 | |||
AK034918.1 | - | 1 | 1196 | |||
AK053145.1 | - | 9 | 1313 | |||
AK144573.1 | - | 25 | 4441 | |||
AK220505.1 | - | 25 | 3079 | |||
BC059267.1 | - | 9 | 872 | |||
BC079911.1 | - | 25 | 2608 | |||
BC145487.1 | - | 26 | 2863 | 9054050 | ||
BC145488.1 | - | 25 | 2839 | 9054051 | ||
BC150779.1 | - | 26 | 2863 | 9087690 |