Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291227.1 | NP_001278156.1 | 18 | 3647 | ||
NM_001291228.1 | NP_001278157.1 | 17 | 3623 | |||
NM_178804.4 | NP_848919.3 | 17 | 3611 | |||
GenBank mRNA | AF144628.1 | - | 17 | 1972 | ||
AK053145.1 | - | 1 | 484 | |||
AK144573.1 | - | 17 | 3612 | |||
BC079911.1 | - | 17 | 1736 | |||
BC145488.1 | - | 17 | 1967 | 9054051 |