Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011421.2 | NP_035551.1 | 2 | 1161 | ||
GenBank mRNA | AK088147.1 | - | 2 | 1130 | ||
AK138133.1 | - | 2 | 1095 | |||
AK145534.1 | - | 2 | 1130 | |||
AK145702.1 | - | 2 | 1162 | |||
AK148189.1 | - | 2 | 1135 | |||
AK149579.1 | - | 2 | 1130 | |||
AK164167.1 | - | 2 | 1130 | |||
AK164717.1 | - | 2 | 1085 | |||
AK167802.1 | - | 2 | 1084 | |||
AK179790.1 | - | 1 | 195 | |||
AK190981.1 | - | 1 | 195 | |||
BC011304.1 | - | 2 | 1096 | 3964394 | ||
BC034515.1 | - | 2 | 1086 | 4482098 | ||
Z14252.1 | - | 2 | 1107 |