Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113559.2 | NP_001107031.2 | 11 | 1575 | ||
NM_001243163.1 | NP_001230092.1 | 11 | 1467 | |||
NM_011444.3 | NP_035574.2 | 12 | 1719 | |||
GenBank mRNA | AB006330.1 | - | 12 | 1631 | ||
AJ010604.1 | - | 11 | 1457 | |||
AK015212.1 | - | 5 | 528 | |||
AK029047.1 | - | 14 | 1717 | |||
AK160154.1 | - | 5 | 533 | |||
BC110478.2 | - | 11 | 1459 | 40047865 | ||
KF793916.1 | - | 12 | 1420 | |||
X65657.1 | - | 5 | 504 |