Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025559.3 | NP_001020730.1 | 16 | 2391 | ||
NM_001025560.2 | NP_001020731.1 | 16 | 2505 | |||
NM_001277326.1 | NP_001264255.1 | 16 | 2486 | |||
NM_001277327.1 | NP_001264256.1 | 15 | 2546 | |||
NM_001277328.1 | NP_001264257.1 | 14 | 2251 | |||
NM_011445.4 | NP_035575.1 | 17 | 2539 | |||
GenBank mRNA | AJ010605.1 | - | 5 | 818 | ||
AK029696.1 | - | 14 | 2179 | |||
AK030713.1 | - | 16 | 2506 | |||
AK132409.1 | - | 16 | 2452 | |||
BC067407.1 | - | 15 | 2543 | 30094413 | ||
D61689.1 | - | 14 | 2247 | |||
U32614.1 | - | 17 | 2543 |