Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079875.2 | NP_001073344.1 | 3 | 653 | ||
NM_009241.3 | NP_033267.2 | 2 | 647 | |||
GenBank mRNA | AK005638.1 | - | 3 | 640 | ||
AK015325.1 | - | 3 | 627 | |||
AY228460.1 | - | 1 | 328 | |||
AY920282.1 | - | 2 | 647 | |||
AY920283.1 | - | 3 | 626 | |||
U33958.1 | - | 2 | 637 |