Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144785.2 | NP_659034.1 | 5 | 1031 | ||
GenBank mRNA | AF529221.1 | - | 5 | 960 | ||
AK052752.1 | - | 5 | 1001 | |||
BC021384.1 | - | 5 | 997 | 5064542 |