Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146010.1 | NP_001139482.1 | 18 | 2262 | ||
NM_199012.2 | NP_950177.2 | 19 | 2334 | |||
GenBank mRNA | AK122368.1 | - | 19 | 2226 | ||
AK140322.1 | - | 18 | 2241 | |||
AK141949.1 | - | 18 | 2262 | |||
BC034086.1 | - | 6 | 808 | |||
BC039788.1 | - | 5 | 736 | |||
BC060647.1 | - | 19 | 2296 | 6854041 |