Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172255.3 | NP_758459.2 | 8 | 1201 | ||
GenBank mRNA | AK043051.1 | - | 6 | 819 | ||
AK166369.1 | - | 8 | 1158 | |||
AK170058.1 | - | 8 | 1149 | |||
AK206398.1 | - | 2 | 138 | |||
BC037177.1 | - | 8 | 1138 | 4948388 | ||
BC042568.1 | - | 1 | 40 |