Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204201.1 | NP_001191130.1 | 8 | 760 | ||
NM_001204202.1 | NP_001191131.1 | 7 | 757 | |||
NM_001204203.1 | NP_001191132.1 | 8 | 708 | |||
NM_001204233.1 | NP_001191162.1 | 6 | 802 | |||
NM_009263.3 | NP_033289.2 | 7 | 705 | |||
GenBank mRNA | AF515708.1 | - | 6 | 605 | ||
AK133555.1 | - | 7 | 696 | |||
AK149588.1 | - | 7 | 696 | |||
AK150819.1 | - | 7 | 696 | |||
AK150845.1 | - | 7 | 696 | |||
AK151069.1 | - | 7 | 666 | |||
AK153089.1 | - | 7 | 695 | |||
AK154908.1 | - | 7 | 696 | |||
AK160540.1 | - | 7 | 699 | |||
AK165393.1 | - | 7 | 696 | |||
AK165965.1 | - | 7 | 748 | |||
AK171158.1 | - | 7 | 696 | |||
BC002113.1 | - | 7 | 702 | 3489075 | ||
BC014284.1 | - | 2 | 364 | |||
BC020355.1 | - | 7 | 685 | |||
BC029991.1 | - | 7 | 685 | |||
BC057858.1 | - | 7 | 678 | 5294167 | ||
BC080720.1 | - | 7 | 678 | 5252869 | ||
CT010357.1 | - | 6 | 605 | |||
J04806.1 | - | 7 | 674 | |||
S78177.1 | - | 5 | 560 | |||
X13986.1 | - | 6 | 620 | |||
X16151.1 | - | 6 | 827 |