Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009266.3 | NP_033292.2 | 1 | 1102 | ||
GenBank mRNA | AK077573.1 | - | 1 | 1103 | ||
AK149516.1 | - | 1 | 1099 | |||
AK168722.1 | - | 1 | 1103 | |||
AK168965.1 | - | 1 | 1103 | |||
BC016508.1 | - | 1 | 1094 | 4911695 | ||
BC028966.1 | - | 1 | 1079 | 5374184 | ||
BC043334.1 | - | 1 | 1083 | 5100207 | ||
U43285.1 | - | 1 | 1099 |