Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Panx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172454.2 | NP_766042.2 | 3 | 689 | ||
GenBank mRNA | AK028288.1 | - | 3 | 656 | ||
AK036933.1 | - | 3 | 645 | |||
BC052822.1 | - | 3 | 657 | 30058169 |