Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160018.1 | NP_001153490.1 | 10 | 1219 | ||
NM_001160019.1 | NP_001153491.1 | 10 | 633 | |||
NM_144791.2 | NP_659040.2 | 7 | 1051 | |||
GenBank mRNA | AB251963.1 | - | 11 | 1216 | ||
AK149727.1 | - | 10 | 1410 | |||
AK152751.1 | - | 10 | 653 | |||
BC010841.1 | - | 7 | 866 | 3493883 | ||
CA465752.1 | - | 10 | 636 |