Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dot1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199322.1 | NP_955354.1 | 7 | 856 | ||
GenBank mRNA | AK035144.1 | - | 7 | 857 | ||
AY196089.1 | - | 7 | 856 | |||
AY376663.1 | - | 5 | 715 | |||
JF508521.1 | - | 7 | 618 |