Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009281.3 | NP_033307.2 | 12, 13 | 1460 | ||
GenBank mRNA | AF011758.2 | - | 1409 | |||
AK028056.1 | - | 12, 13 | 1453 | |||
AK029078.1 | - | 12, 13 | 1452 | |||
AK029926.1 | - | 12, 13 | 1725 | |||
BC037658.1 | - | 11, 12 | 1370 | 5356405 |