Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 5 | 578 | ||
GenBank mRNA | AF088862.1 | - | 5 | 448 | ||
AF187231.1 | - | 5 | 572 | |||
AF206162.1 | - | 5 | 578 | |||
AK149838.1 | - | 5 | 575 | |||
AK149924.1 | - | 5 | 591 | |||
AK150070.1 | - | 5 | 590 | |||
AK153405.1 | - | 3 | 216 |