Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 11, 12 | 1242 | ||
GenBank mRNA | AF187231.1 | - | 11, 12 | 1236 | ||
AF206162.1 | - | 11, 12 | 1242 | |||
AK030903.1 | - | 4, 5 | 334 | |||
AK149924.1 | - | 11, 12 | 1255 | |||
AK150070.1 | - | 11, 12 | 1254 | |||
AK153405.1 | - | 9, 10 | 880 |