Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109905.2 | NP_001103375.1 | 7 | 1050 | ||
NM_001109906.2 | NP_001103376.1 | 7 | 1068 | |||
NM_001291160.1 | NP_001278089.1 | 7 | 1050 | |||
NM_011490.4 | NP_035620.1 | 7 | 1044 | |||
GenBank mRNA | AF061942.1 | - | 7 | 1044 | ||
AF395842.1 | - | 6 | 771 | |||
AK005002.2 | - | 7 | 991 | |||
AK139532.1 | - | 7 | 997 | |||
AK145826.1 | - | 7 | 1035 | |||
AK151835.1 | - | 7 | 856 | |||
AK152269.1 | - | 7 | 862 | |||
AK152744.1 | - | 7 | 856 | |||
AK158952.1 | - | 3 | 652 | |||
AK159602.1 | - | 7 | 985 | |||
AK159781.1 | - | 7 | 1020 | |||
AK160979.1 | - | 7 | 1009 | |||
AK167637.1 | - | 7 | 991 | |||
AK194004.1 | - | 2 | 47 | |||
AK206904.1 | - | 2 | 47 | |||
AK210669.1 | - | 2 | 47 | |||
AY391773.1 | - | 6 | 801 | |||
BC012959.1 | - | 7 | 1014 | 4188394 | ||
BC082277.1 | - | 7 | 987 |