Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009285.3 | NP_033311.3 | 3 | 409 | ||
GenBank mRNA | AF099098.1 | - | 3 | 461 | ||
AK134260.1 | - | 3 | 410 | |||
AK134331.1 | - | 1 | 57 | |||
AK142220.1 | - | 3 | 308 | |||
AK149293.1 | - | 3 | 292 | |||
AK149351.1 | - | 3 | 292 | |||
AK161960.1 | - | 3 | 291 | |||
BC021425.1 | - | 3 | 430 | 5010981 |