Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kntc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042421.1 | NP_001035886.1 | 41 | 4186 | ||
GenBank mRNA | AK172905.1 | - | 41 | 4160 | ||
BC023686.1 | - | 8 | 916 |