Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016737.2 | NP_058017.1 | 4 | 597 | ||
GenBank mRNA | AK075988.1 | - | 4 | 535 | ||
AK088494.1 | - | 4 | 539 | |||
AK134171.1 | - | 4 | 513 | |||
AK149493.1 | - | 4 | 513 | |||
AK161645.1 | - | 4 | 516 | |||
AK167273.1 | - | 4 | 531 | |||
AK168181.1 | - | 4 | 513 | |||
BC003794.1 | - | 4 | 506 | 3590457 | ||
U27830.1 | - | 4 | 514 |