Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277992.1 | NP_001264921.1 | 3 | 407 | ||
NM_011494.5 | NP_035624.3 | 3 | 435 | |||
GenBank mRNA | AB017197.1 | - | 3 | 383 | ||
AB022179.1 | - | 3 | 365 | |||
AF062076.1 | - | 2 | 122 | |||
AF089869.1 | - | 3 | 412 | |||
AJ005790.1 | - | 3 | 366 | |||
AK030404.1 | - | 3 | 410 | |||
AK032162.1 | - | 3 | 426 | |||
AK149539.1 | - | 3 | 408 | |||
AK156554.1 | - | 3 | 338 | |||
AK179125.1 | - | 2 | 95 | |||
AK180891.1 | - | 2 | 95 | |||
AK181414.1 | - | 2 | 95 | |||
AK182583.1 | - | 2 | 95 | |||
BC005703.1 | - | 3 | 394 | 3598470 | ||
BC028999.1 | - | 3 | 394 | 5250877 | ||
CX200029.1 | - | 3 | 382 |