Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277992.1 | NP_001264921.1 | 3 | 519 | ||
NM_011494.5 | NP_035624.3 | 3 | 547 | |||
GenBank mRNA | AA183397.1 | - | 1 | 16 | ||
AB017197.1 | - | 3 | 495 | |||
AB022179.1 | - | 3 | 477 | |||
AF062076.1 | - | 2 | 234 | |||
AF089869.1 | - | 3 | 524 | |||
AJ005790.1 | - | 3 | 478 | |||
AK030404.1 | - | 3 | 522 | |||
AK032162.1 | - | 3 | 538 | |||
AK149539.1 | - | 3 | 520 | |||
AK156554.1 | - | 3 | 450 | |||
BC005703.1 | - | 3 | 506 | 3598470 | ||
BC028999.1 | - | 3 | 506 | 5250877 | ||
CX200029.1 | - | 3 | 494 |