Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145943.1 | NP_666055.1 | 7 | 1325 | ||
GenBank mRNA | AK138767.1 | - | 7 | 1322 | ||
AK160893.1 | - | 7 | 1348 | |||
BC031781.1 | - | 7 | 1325 | 3672316 |