Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290826.1 | NP_001277755.1 | 6 | 1903 | ||
NM_001290827.1 | NP_001277756.1 | 6 | 1552 | |||
NM_001290828.1 | NP_001277757.1 | 6 | 1545 | |||
NM_001290829.1 | NP_001277758.1 | 5 | 1402 | |||
NM_198300.3 | NP_938042.2 | 6 | 1614 | |||
GenBank mRNA | AB093274.1 | - | 6 | 1543 | ||
AK029261.1 | - | 5 | 1403 | |||
AK044639.1 | - | 6 | 1547 | |||
AK147243.1 | - | 6 | 1904 | |||
AK161513.1 | - | 6 | 1521 | |||
AY313774.1 | - | 6 | 1436 | |||
BC128377.1 | - | 7 | 959 | 40111837 |