Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhlhe40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011498.4 | NP_035628.1 | 2, 3 | 428 | ||
GenBank mRNA | AF010305.1 | - | 2, 3 | 360 | ||
AF364051.1 | - | 2, 3 | 360 | |||
AK048242.1 | - | 2, 3 | 429 | |||
AK090032.1 | - | 2, 3 | 382 | |||
AK154935.1 | - | 2, 3 | 381 | |||
BC010720.1 | - | 2, 3 | 371 | 3707474 | ||
Y07836.1 | - | 2, 3 | 123 |