Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162475.1 | NP_001155947.1 | 15 | 1439 | ||
NM_001162476.1 | NP_001155948.1 | 15 | 1750 | |||
NM_001162479.1 | NP_001155951.1 | 15 | 1544 | |||
NM_009291.2 | NP_033317.2 | 15 | 1709 | |||
GenBank mRNA | AF062476.1 | - | 15 | 1709 | ||
BC075657.1 | - | 15 | 1439 | 30605532 | ||
CF735552.1 | - | 7 | 609 |