Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 12, 13 | 1365 | ||
NM_001285894.1 | NP_001272823.1 | 13, 14 | 1318 | |||
GenBank mRNA | AK122375.1 | - | 13, 14 | 1267 | ||
AK141678.1 | - | 12, 13 | 1288 | |||
BC055323.1 | - | 12, 13 | 1282 | 5706288 |