Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011505.2 | NP_035635.1 | 13 | 1398 | ||
GenBank mRNA | AF152924.1 | - | 13 | 1388 | ||
AK039029.1 | - | 1 | 72 | |||
AK138499.1 | - | 1 | 71 | |||
BC032881.1 | - | 13 | 1352 | 1224878 |