Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009297.2 | NP_033323.2 | 26, 27 | 3671 | ||
GenBank mRNA | AK129069.1 | - | 14, 15 | 1994 | ||
AK158846.1 | - | 8, 9 | 1606 | |||
AK170533.1 | - | 26, 27 | 3670 | |||
BC072657.1 | - | 26, 27 | 3591 | 30536338 | ||
U40375.1 | - | 26, 27 | 3669 |