Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271724.1 | NP_001258653.2 | 5 | 675 | ||
NM_013677.2 | NP_038705.2 | 5 | 675 | |||
GenBank mRNA | AK002469.1 | - | 5 | 489 | ||
AK154726.1 | - | 5 | 495 | |||
AK208539.1 | - | 1 | 18 | |||
BC004755.1 | - | 5 | 675 | 3584373 | ||
BC052500.1 | - | 5 | 662 | 5255619 |