Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011512.3 | NP_035642.1 | 6 | 985 | ||
GenBank mRNA | AK004691.2 | - | 6 | 900 | ||
AK128902.1 | - | 6 | 899 | |||
AK145670.1 | - | 6 | 926 | |||
AK150568.1 | - | 6 | 803 | |||
AK151161.1 | - | 6 | 855 | |||
AK151319.1 | - | 6 | 813 | |||
AK152692.1 | - | 6 | 895 | |||
AK190206.1 | - | 1 | 120 | |||
AK190383.1 | - | 1 | 120 | |||
AK197835.1 | - | 1 | 120 | |||
AK204224.1 | - | 1 | 120 | |||
AK207760.1 | - | 1 | 120 | |||
AK219081.1 | - | 1 | 120 | |||
BC003280.1 | - | 6 | 884 | 3582114 | ||
BC027352.1 | - | 6 | 845 | 3594564 | ||
M63114.1 | - | 6 | 1401 |