Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172473.3 | NP_766061.2 | 20 | 2470 | ||
GenBank mRNA | AK029162.1 | - | 10 | 1290 | ||
AK042879.1 | - | 20 | 2411 | |||
AK156958.1 | - | 20 | 2412 | |||
AK220383.1 | - | 2 | 160 | |||
BC120695.1 | - | 20 | 2367 | 40129618 | ||
BC120697.1 | - | 20 | 2367 | 40129620 |