Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145952.3 | NP_666064.3 | 8 | 1548 | ||
GenBank mRNA | AK172979.1 | - | 8 | 1542 | ||
BC138056.1 | - | 8 | 1444 | 8861075 | ||
BC138057.1 | - | 8 | 1444 | 8861076 | ||
BC145129.1 | - | 6 | 1219 | 9053677 |