Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173748.4 | NP_776109.1 | 5 | 1174 | ||
GenBank mRNA | AK030050.1 | - | 5 | 900 | ||
AK036694.1 | - | 5 | 879 | |||
AK049739.1 | - | 5 | 886 | |||
AK050476.1 | - | 4 | 720 | |||
AK159391.1 | - | 5 | 906 | |||
AK159458.1 | - | 5 | 906 | |||
AK169797.1 | - | 5 | 910 | |||
AK170329.1 | - | 5 | 907 | |||
AK171224.1 | - | 5 | 889 | |||
AK204837.1 | - | 2 | 81 | |||
BC024322.1 | - | 5 | 892 | 4236345 | ||
BC037090.1 | - | 5 | 873 | 5134044 | ||
BC052089.1 | - | 5 | 735 | |||
BC057603.1 | - | 4 | 642 | |||
EU350933.1 | - | 5 | 865 |