Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il23r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144548.1 | NP_653131.2 | 11 | 1933 | ||
GenBank mRNA | AF461423.1 | - | 11 | 1933 | ||
BC112425.1 | - | 11 | 1916 | 40062206 |