Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172477.4 | NP_766065.1 | 6 | 1828 | ||
GenBank mRNA | AK048041.1 | - | 6 | 1813 | ||
AK081176.1 | - | 6 | 1829 | |||
AK135781.1 | - | 4 | 373 | |||
AK160784.1 | - | 6 | 1590 | |||
AK165202.1 | - | 6 | 1684 | |||
AK167298.1 | - | 6 | 1682 | |||
BC056367.1 | - | 6 | 1698 | |||
BC057310.1 | - | 6 | 1698 | |||
BC145599.1 | - | 5 | 1507 | 9054165 | ||
BC150799.1 | - | 5 | 1507 | 9087710 |