Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024138.1 | NP_001019309.1 | 2 | 513 | ||
GenBank mRNA | AB196532.1 | - | 2 | 513 | ||
AY485344.1 | - | 2 | 513 | |||
BC125496.1 | - | 2 | 587 | 40130011 | ||
BC125498.1 | - | 2 | 587 | 40130013 |