Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171824.2 | NP_741958.1 | 3, 4 | 704 | ||
GenBank mRNA | BC094384.1 | - | 702 | 30606749 | ||
CA316156.1 | - | 2, 3 | 507 |