Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153534.2 | NP_705762.2 | 9 | 1604 | ||
GenBank mRNA | AK122432.1 | - | 10 | 1419 | ||
AK132312.1 | - | 9 | 1605 | |||
AK197076.1 | - | 1 | 77 | |||
BC037107.1 | - | 9 | 1361 | 5367175 |