Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145952.1 | NP_001139424.1 | 7 | 997 | ||
NM_001145954.1 | NP_001139426.1 | 3 | 411 | |||
NM_178665.5 | NP_848780.3 | 8 | 1204 | |||
GenBank mRNA | AK029335.1 | - | 8 | 996 | ||
AK029567.1 | - | 3 | 412 | |||
AK040643.1 | - | 6 | 758 | |||
AK045288.1 | - | 8 | 1065 | |||
AK051345.1 | - | 8 | 1076 | |||
AK054550.1 | - | 7 | 1000 | |||
AK076989.1 | - | 3 | 402 | |||
AK085626.1 | - | 8 | 1196 | |||
AK086307.1 | - | 8 | 1015 | |||
AK168431.1 | - | 8 | 1111 | |||
AK169780.1 | - | 8 | 1016 | |||
BC085321.1 | - | 7 | 943 | 30743832 |