Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161366.1 | NP_001154838.1 | 1 | 5557 | ||
NM_001161367.1 | NP_001154839.1 | 1 | 5557 | |||
NM_198418.2 | NP_940810.2 | 1 | 5557 | |||
GenBank mRNA | AB097085.1 | - | 1 | 5557 | ||
BC138353.1 | - | 2 | 4945 | 8861373 | ||
BC145247.1 | - | 1 | 5523 | |||
BC145248.1 | - | 1 | 5523 | 9053800 |