Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB097085.1 | - | 1 | 4838 | ||
BC138353.1 | - | 2 | 4226 | 8861373 | ||
BC145247.1 | - | 1 | 4804 | |||
BC145248.1 | - | 1 | 4804 | 9053800 |