Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201638.2 | NP_964000.2 | 8 | 840 | ||
GenBank mRNA | AK083169.1 | - | 7 | 1464 | ||
AK129409.1 | - | 798 | ||||
AK146881.1 | - | 8 | 829 | |||
AK153810.1 | - | 8 | 836 | |||
BC052204.1 | - | 8 | 798 | 30007932 |