Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181584.4 | NP_853615.1 | 5, 6 | 1288 | ||
GenBank mRNA | AJ748654.1 | - | 2, 3 | 135 | ||
AK031227.1 | - | 5, 6 | 1286 | |||
AY057988.1 | - | 5, 6 | 1174 | |||
BB634123.1 | - | 2, 3 | 283 | |||
BC141240.1 | - | 5, 6 | 1168 | 9055852 |