Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177595.4 | NP_808263.2 | 5 | 909 | ||
GenBank mRNA | AK034675.1 | - | 5 | 887 | ||
BC137728.1 | - | 5 | 844 | 8860747 |