Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 2 | 393 | ||
NM_153414.4 | NP_700463.2 | 2 | 361 | |||
GenBank mRNA | AK035769.1 | - | 2 | 105 | ||
AK038979.1 | - | 2 | 94 | |||
AK077634.1 | - | 2 | 362 | |||
AK136992.1 | - | 2 | 88 | |||
AK150195.1 | - | 2 | 88 | |||
BC028953.1 | - | 1 | 51 | 5362016 | ||
BC055700.1 | - | 2 | 60 | 6808816 | ||
BY195822.1 | - | 2 | 126 |