Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178591.2 | NP_848706.2 | 7 | 1189 | ||
GenBank mRNA | AK170314.1 | - | 7 | 697 | ||
AY648975.1 | - | 7 | 1189 | |||
AY648976.1 | - | 11 | 1024 | |||
BC151113.1 | - | 7 | 1273 | 9088026 |